Thermo Scientific Chemicals

3-Methoxythiophenol, 97%, Thermo Scientific Chemicals

Catalog number: A10648.03
1 g, Each
Thermo Scientific Chemicals

3-Methoxythiophenol, 97%, Thermo Scientific Chemicals

Catalog number: A10648.03
1 g, Each
Quantity
Catalog number: A10648.03
also known as A10648-03
Price (USD)
Price: 28.50
Online price: 24.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15570-12-4
IUPAC Name
3-methoxybenzene-1-thiol
Molecular Formula
C7H8OS
InChI Key
QMVAZEHZOPDGHA-UHFFFAOYSA-N
SMILES
COC1=CC=CC(S)=C1
Form
Liquid
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.5850-1.5900 @ 20?C

Description

3-Methoxythiophenol is used in the preparation of allenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxythiophenol is used in the preparation of allenes.

Solubility
Not miscible or difficult to mix in water. Soluble in methanol, benzene, hexane, toluene and dichloromethane.

Notes
Store under dry inert gas. It is sensitive to air. Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text