Thermo Scientific Chemicals

2-Chloro-5-nitropyridine, 99%, Thermo Scientific Chemicals

Catalog number: A10709.22
100 g, Each
Thermo Scientific Chemicals

2-Chloro-5-nitropyridine, 99%, Thermo Scientific Chemicals

Catalog number: A10709.22
100 g, Each
Quantity
Catalog number: A10709.22
also known as A10709-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4548-45-2
IUPAC Name
2-chloro-5-nitropyridine
Molecular Formula
C5H3ClN2O2
InChI Key
BAZVFQBTJPBRTJ-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(Cl)N=C1
Form
crystals or crystalline powder
Solution Test
0.2g in 20ml methanol. Matches Sol 'A' or 'B' toJIS K8001 5.2
Assay (GC)
>98.5%
Appearance (Color)
Pale yellow or cream to pale brown

Description

2-Chloro-5-nitropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-nitropyridine is used as a pharmaceutical intermediate.

Solubility
Solubility in hot methanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text