Thermo Scientific Chemicals

1-Bromopentane, 99%, Thermo Scientific Chemicals

Catalog number: A10821.0I
5000 g, Each
Thermo Scientific Chemicals

1-Bromopentane, 99%, Thermo Scientific Chemicals

Catalog number: A10821.0I
5000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10821.0I
also known as A10821-0I
Price (USD)
Price: 483.00
Online price: 410.65
Your price:
Quantity
-

Chemical Identifiers

CAS
110-53-2
IUPAC Name
1-bromopentane
Molecular Formula
C5H11Br
InChI Key
YZWKKMVJZFACSU-UHFFFAOYSA-N
SMILES
CCCCCBr
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4420-1.4460 @ 20?C
Assay (GC)
≥98.5%

Description

1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.

Solubility
Insoluble in water and soluble in alcohol. Miscible in diethyl ether.

Notes
Store away from oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text