Thermo Scientific Chemicals

Pyridine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A10871.30
250 g, Each
Thermo Scientific Chemicals

Pyridine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A10871.30
250 g, Each
Quantity
Catalog number: A10871.30
also known as A10871-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
628-13-7
IUPAC Name
pyridin-1-ium chloride
Molecular Formula
C5H6ClN
InChI Key
AOJFQRQNPXYVLM-UHFFFAOYSA-N
SMILES
[Cl-].C1=CC=[NH+]C=C1
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder or lumps or fused solid
Water Content (Karl Fischer Titration)
≤1.5%
Assay (Freebase GC)
≥97.5%

Description

Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrochloride is a classical reagent for cleavage of aryl methyl ethers at 200-220o. It is also used in the demethylation of 4,5-dimethyl-7-methoxy-1-tetralone.

Solubility
Soluble in water (85 g/100 ml) and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text