Thermo Scientific Chemicals

Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10926.30
250 g, Each
Thermo Scientific Chemicals

Diethylenetriaminepentaacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10926.30
250 g, Each
Quantity
Catalog number: A10926.30
also known as A10926-30
Price (USD)
Price: 51.40
Online price: 43.65
Your price:
Quantity
-

Chemical Identifiers

CAS
67-43-6
IUPAC Name
2-[bis({2-[bis(carboxymethyl)amino]ethyl})amino]acetic acid
Molecular Formula
C14H23N3O10
InChI Key
QPCDCPDFJACHGM-UHFFFAOYSA-N
SMILES
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O
Assay (unspecified)
≥98.0%
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White

Description

Diethylenetriaminepentaacetic acid is used as chelating agent and forms complexes with metal ions. It finds application as a magnetic resonance imaging (MRI) contrasting agent, since it improves the image by forming a complex with a gadolinium ion. It can also be used for treatment of radioactive materials such as plutonium, americium and other actinides. It is an active component of manganese and zinc fertilizers used in agrochemical industry. It acts as a color inhibitor and is used in the production of acrylon. It is also used in soaps as a water softener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylenetriaminepentaacetic acid is used as chelating agent and forms complexes with metal ions. It finds application as a magnetic resonance imaging (MRI) contrasting agent, since it improves the image by forming a complex with a gadolinium ion. It can also be used for treatment of radioactive materials such as plutonium, americium and other actinides. It is an active component of manganese and zinc fertilizers used in agrochemical industry. It acts as a color inhibitor and is used in the production of acrylon. It is also used in soaps as a water softener.

Solubility
Soluble in water and alkali.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text