Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-tyrosine, 99%, may contain up to ca 10% water, Thermo Scientific Chemicals

Catalog number: A10966.06
5 g, Each
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-tyrosine, 99%, may contain up to ca 10% water, Thermo Scientific Chemicals

Catalog number: A10966.06
5 g, Each
Quantity
Catalog number: A10966.06
also known as A10966-06
Price (USD)

Chemical Identifiers

CAS
1164-16-5
IUPAC Name
(2S)-2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Molecular Formula
C17H17NO5
InChI Key
MCRMUCXATQAAMN-GGYSOQFKNA-N
SMILES
OC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (dry weight basis, non-U.S. specification)
Melting Point (clear melt)
55-60°C (non-U.S. specification)
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text