Thermo Scientific Chemicals

Dimethyl malonate, 98+%, Thermo Scientific Chemicals

Catalog number: A11007.36
500 g, Each
Thermo Scientific Chemicals

Dimethyl malonate, 98+%, Thermo Scientific Chemicals

Catalog number: A11007.36
500 g, Each
Quantity
Catalog number: A11007.36
also known as A11007-36
Price (USD)
Price: 53.20
Online price: 45.65
Your price:
Quantity
-

Chemical Identifiers

CAS
108-59-8
IUPAC Name
1,3-dimethyl propanedioate
Molecular Formula
C5H8O4
InChI Key
BEPAFCGSDWSTEL-UHFFFAOYSA-N
SMILES
COC(=O)CC(=O)OC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4115-1.4155 @ 20?C

Description

Dimethyl malonate is a reagent used in organic synthesis. It acts as a precursor for the synthesis of mono-substituted and di-substituted acetic acid, barbiturates, vitamin B1 and vitamin B6. It is used in the pharmaceutical industry to prepare pharmaceuticals like chloroquine and butazolidin. It is involved in the synthesis of diastereomeric pure thienylpyridines by reacting with 1,3-diphenylprop-2-enyl acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl malonate is a reagent used in organic synthesis. It acts as a precursor for the synthesis of mono-substituted and di-substituted acetic acid, barbiturates, vitamin B1 and vitamin B6. It is used in the pharmaceutical industry to prepare pharmaceuticals like chloroquine and butazolidin. It is involved in the synthesis of diastereomeric pure thienylpyridines by reacting with 1,3-diphenylprop-2-enyl acetate.

Solubility
Miscible with alcohol, ether and oils. Slightly miscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text