Ethyl 4-nitrophenylacetate, 98%
Ethyl 4-nitrophenylacetate, 98%
Ethyl 4-nitrophenylacetate, 98%
Thermo Scientific Chemicals

Ethyl 4-nitrophenylacetate, 98%

CAS: 5445-26-1 | C10H11NO4 | 209.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A11101.06
also known as A11101-06
Price (USD)
55.65
Online Exclusive
61.50
Save 5.85 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.50
Save 5.85 (10%)
Each
Chemical Identifiers
CAS5445-26-1
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream or pale yellow
Assay (GC)≥97.5%
Melting Point (clear melt)59.5-65.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only