Thermo Scientific Chemicals

Carbohydrazide, 97%, Thermo Scientific Chemicals

Catalog number: A11145.36
500 g, Each
Thermo Scientific Chemicals

Carbohydrazide, 97%, Thermo Scientific Chemicals

Catalog number: A11145.36
500 g, Each
Quantity
Catalog number: A11145.36
also known as A11145-36
Price (USD)
Price: 606.00
Online price: 515.65
Your price:
Quantity
-

Chemical Identifiers

CAS
497-18-7
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Melting Point (clear melt)
150.0-160.0?C
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

Carbohydrazide is widely utilized as a curing agent for epoxide-type resins and as an oxygen scrubber in boiler systems. It is applied in photography to stabilize color developers that produce images of the azo-methine and azine classes. It is used to develop ammunition propellants and stabilize soaps. It is also a useful reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbohydrazide is widely utilized as a curing agent for epoxide-type resins and as an oxygen scrubber in boiler systems. It is applied in photography to stabilize color developers that produce images of the azo-methine and azine classes. It is used to develop ammunition propellants and stabilize soaps. It is also a useful reagent in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, copper, zinc, nickel, lead and brass.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text