Thermo Scientific Chemicals

3-Methyl-1-phenyl-2-pyrazolin-5-one, 98+%, Thermo Scientific Chemicals

Catalog number: A11161.22
100 g, Each
Thermo Scientific Chemicals

3-Methyl-1-phenyl-2-pyrazolin-5-one, 98+%, Thermo Scientific Chemicals

Catalog number: A11161.22
100 g, Each
Quantity
Catalog number: A11161.22
also known as A11161-22
Price (USD)
Price: 26.10
Online price: 22.65
Your price:
Quantity
-

Chemical Identifiers

CAS
89-25-8
IUPAC Name
3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Molecular Formula
C10H10N2O
InChI Key
QELUYTUMUWHWMC-UHFFFAOYSA-N
SMILES
CC1=NN(C(=O)C1)C1=CC=CC=C1
Comment
Material sourced in the U.S. and in other countries
Melting Point (clear melt)
124.0-133.0°C
Appearance (Color)
Pale cream to cream to yellow to pale brown
Form
Powder
Assay (GC)
≥98.0%

Description

3-Methyl-1-phenyl-2-pyrazolin-5-one used as reagent for detection of reducing carbohydrates by ESI/MALDI -MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-pyrazolin-5-one used as reagent for detection of reducing carbohydrates by ESI/MALDI -MS.

Solubility
Soluble in water(3.3g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text