Thermo Scientific Chemicals

3-Methyl-4-nitrobenzonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A11301.09
10 g, Each
Thermo Scientific Chemicals

3-Methyl-4-nitrobenzonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A11301.09
10 g, Each
Quantity
Catalog number: A11301.09
also known as A11301-09
Price (USD)
Price: 182.00
Online price: 156.65
Your price:
Quantity
-

Chemical Identifiers

CAS
96784-54-2
IUPAC Name
3-methyl-4-nitrobenzonitrile
Molecular Formula
C8H6N2O2
InChI Key
IHVNKSXTJZNBQA-UHFFFAOYSA-N
SMILES
CC1=CC(=CC=C1[N+]([O-])=O)C#N
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
72.5-81.5°C

Description

3-Methyl-4-nitrobenzonitrile is used to produce 4-amino-3-methyl-benzonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitrobenzonitrile is used to produce 4-amino-3-methyl-benzonitrile.

Solubility
Insoluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text