Thermo Scientific Chemicals

Sodium dihydrogen phosphate dihydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A11316.22
100 g, Each
Thermo Scientific Chemicals

Sodium dihydrogen phosphate dihydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A11316.22
100 g, Each
Quantity
Catalog number: A11316.22
also known as A11316-22
Price (USD)
Price: 31.60
Online price: 26.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13472-35-0
IUPAC Name
sodium dihydrate dihydrogen phosphate
Molecular Formula
H6NaO6P
InChI Key
VBJGJHBYWREJQD-UHFFFAOYSA-M
SMILES
O.O.[Na+].OP(O)([O-])=O
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

Solubility
Soluble in water.(1 M at 20°C)

Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text