Thermo Scientific Chemicals

Butyrophenone, 99%, Thermo Scientific Chemicals

Catalog number: A11345.30
250 g, Each
Thermo Scientific Chemicals

Butyrophenone, 99%, Thermo Scientific Chemicals

Catalog number: A11345.30
250 g, Each
Quantity
Catalog number: A11345.30
also known as A11345-30
Price (USD)
Price: 210.00
Online price: 180.65
Your price:
Quantity
-

Chemical Identifiers

CAS
495-40-9
IUPAC Name
1-phenylbutan-1-one
Molecular Formula
C10H12O
InChI Key
FFSAXUULYPJSKH-UHFFFAOYSA-N
SMILES
CCCC(=O)C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.5%

Description

Butyrophenone is used as intermediates of Liquid Crystals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyrophenone is used as intermediates of Liquid Crystals

Solubility
Insoluble in water.

Notes
Protect from light. Store under an inert atmosphere.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text