Thermo Scientific Chemicals

6-Fluoro-2-methylquinoline, 98+%, Thermo Scientific Chemicals

Catalog number: A11469.22
100 g, Each
Thermo Scientific Chemicals

6-Fluoro-2-methylquinoline, 98+%, Thermo Scientific Chemicals

Catalog number: A11469.22
100 g, Each
Quantity
Catalog number: A11469.22
also known as A11469-22
Price (USD)
Price: 1,073.00
Online price: 927.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1128-61-6
IUPAC Name
6-fluoro-2-methylquinoline
Molecular Formula
C10H8FN
InChI Key
GPIARMSVZOEZCV-UHFFFAOYSA-N
SMILES
CC1=CC=C2C=C(F)C=CC2=N1
Appearance (Color)
White to pale cream to pale brown to gray
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
49.0-55.0?C

Description

Metal-free transfer hydrogenation of 6-fluoro-2-methylquinoline by employing several chiral Bronsted acid catalysts was carried out in combination with dihydropyridines as the hydride source in asymmetric metal-free synthesis of fluoroquinolones by organocatalytic hydrogenation. Hydrogenation product of 6-fluoro-2-methylquinoline is the key intermediate of the antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metal-free transfer hydrogenation of 6-fluoro-2-methylquinoline by employing several chiral Brønsted acid catalysts was carried out in combination with dihydropyridines as the hydride source in asymmetric metal-free synthesis of fluoroquinolones by organocatalytic hydrogenation. Hydrogenation product of 6-fluoro-2-methylquinoline is the key intermediate of the antibacterial agent.

Solubility
Soluble in Chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text