Thermo Scientific Chemicals

Calcium D-gluconate monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A11649.0E
2500 g, Each
Thermo Scientific Chemicals

Calcium D-gluconate monohydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A11649.0E
2500 g, Each
Quantity
Catalog number: A11649.0E
also known as A11649-0E
Price (USD)
Price: 146.00
Online price: 124.65
Your price:
Quantity
-

Chemical Identifiers

CAS
66905-23-5
IUPAC Name
calcium bis(2,3,4,5,6-pentahydroxyhexanoate) hydrate
Molecular Formula
C12H24CaO15
InChI Key
XLNFVCRGJZBQGX-UHFFFAOYNA-L
SMILES
O.[Ca++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O
Appearance (Color)
White
Chloride content
≤0.08%
Optical Rotation
9.0? ? 1.0? (c=1.2, water)
Loss on Drying
≤1.5% (105?C/constant wt.)
Complexometric Titration
≥98.0 to ≤102.0%

Description

Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.

Solubility
Soluble in water(30g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text