Thermo Scientific Chemicals

9-Ethylcarbazole, 99%, Thermo Scientific Chemicals

Catalog number: A11653.0B
1000 g, Each
Thermo Scientific Chemicals

9-Ethylcarbazole, 99%, Thermo Scientific Chemicals

Catalog number: A11653.0B
1000 g, Each
Quantity
Catalog number: A11653.0B
also known as A11653-0B
Price (USD)
Price: 242.00
Online price: 205.65
Your price:
Quantity
-

Chemical Identifiers

CAS
86-28-2
IUPAC Name
9-ethyl-9H-carbazole
Molecular Formula
C14H13N
InChI Key
PLAZXGNBGZYJSA-UHFFFAOYSA-N
SMILES
CCN1C2=C(C=CC=C2)C2=C1C=CC=C2
Identification (FTIR)
Conforms
Melting Point (clear melt)
66.0-73.0°C
Appearance (Color)
White to cream or brown
Form
Crystals or powder or crystalline powder or flakes or needles
Assay (GC)
≥98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text