Thermo Scientific Chemicals

4-Dimethylaminobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A11712.22
100 g, Each
Thermo Scientific Chemicals

4-Dimethylaminobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A11712.22
100 g, Each
Quantity
Catalog number: A11712.22
also known as A11712-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
100-10-7
IUPAC Name
4-(dimethylamino)benzaldehyde
Molecular Formula
C9H11NO
InChI Key
BGNGWHSBYQYVRX-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=O)C=C1
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or flakes
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to green to pale brown or blue to black
Free acid (titration)
≤1%

Description

4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

Solubility
Soluble in alcohol, ether, chloroform, acetic acid. Insoluble in water.

Notes
Light sensitive. Stable at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text