Thermo Scientific Chemicals

2-Mercaptopyridine, 98%, Thermo Scientific Chemicals

Catalog number: A11741.22
100 g, Each
Thermo Scientific Chemicals

2-Mercaptopyridine, 98%, Thermo Scientific Chemicals

Catalog number: A11741.22
100 g, Each
Quantity
Catalog number: A11741.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2637-34-5
IUPAC Name
1,2-dihydropyridine-2-thione
Molecular Formula
C5H5NS
InChI Key
WHMDPDGBKYUEMW-UHFFFAOYSA-N
SMILES
S=C1NC=CC=C1
Appearance (Color)
Yellow
Assay (HPLC)
≥97.5%
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder

Description

Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

Solubility
Slightly soluble in chloroform, methanol and dimethyl sulfoxide.

Notes
Air Sensitive, keep the container tightly sealed. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text