Thermo Scientific Chemicals

2-Chloro-1-fluoro-4-iodobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A11808.22
100 g, Each
Thermo Scientific Chemicals

2-Chloro-1-fluoro-4-iodobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A11808.22
100 g, Each
Quantity
Catalog number: A11808.22
also known as A11808-22
Price (USD)
Price: 442.00
Online price: 382.65
Your price:
Quantity
-

Chemical Identifiers

CAS
156150-67-3
IUPAC Name
2-chloro-1-fluoro-4-iodobenzene
Molecular Formula
C6H3ClFI
InChI Key
OMASDGWBVAVFQZ-UHFFFAOYSA-N
SMILES
FC1=C(Cl)C=C(I)C=C1
Refractive Index
1.6020-1.6060 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid

Description

It employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text