Thermo Scientific Chemicals

4-Diethylaminobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A11825.22
100 g, Each
Thermo Scientific Chemicals

4-Diethylaminobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A11825.22
100 g, Each
Quantity
Catalog number: A11825.22
also known as A11825-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
120-21-8
IUPAC Name
4-(diethylamino)benzaldehyde
Molecular Formula
C11H15NO
InChI Key
MNFZZNNFORDXSV-UHFFFAOYSA-N
SMILES
CCN(CC)C1=CC=C(C=O)C=C1
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Appearance (Color)
Yellow to dark green to pale brown
Free acid (titration)
≤1.5%
Form
Crystals or powder or crystalline powder or lumps

Description

4-Diethylaminobenzaldehyde is used in the preparation of 5-(4-diethylamino-benzylidene)-selenazolidine-2,4-dione by using aqueous methylamine as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Diethylaminobenzaldehyde is used in the preparation of 5-(4-diethylamino-benzylidene)-selenazolidine-2,4-dione by using aqueous methylamine as a reagent.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text