Thermo Scientific Chemicals

1-Fluoro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A11871.22
100 g, Each
Thermo Scientific Chemicals

1-Fluoro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A11871.22
100 g, Each
Quantity
Catalog number: A11871.22
also known as A11871-22
Price (USD)
Price: 258.00
Online price: 219.65
Your price:
Quantity
-

Chemical Identifiers

CAS
70-34-8
IUPAC Name
1-fluoro-2,4-dinitrobenzene
Molecular Formula
C6H3FN2O4
InChI Key
LOTKRQAVGJMPNV-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
Form
Low melting solid or clear liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5675-1.5705 as melt
Appearance (Color)
Yellow to orange or brown

Description

1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-2,4-dinitrobenzene is used to identify the amino acid sequence. It reacts with amino group of amino acids to yield dinitrophenyl-amino acids. It is also used in chromatographic methods. Further, it acts as an alkylating agent used in elucidating amino acid sequence in proteins.

Solubility
Soluble in chloroform, benzene, ether and propylene glycol.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text