Thermo Scientific Chemicals

Methyl tiglate, 98%, Thermo Scientific Chemicals

Catalog number: A11964.18
50 g, Each
Thermo Scientific Chemicals

Methyl tiglate, 98%, Thermo Scientific Chemicals

Catalog number: A11964.18
50 g, Each
Quantity
Catalog number: A11964.18
also known as A11964-18
Price (USD)
Price: 141.00
Online price: 121.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6622-76-0
IUPAC Name
methyl (2Z)-2-methylbut-2-enoate
Molecular Formula
C6H10O2
InChI Key
YYJWBYNQJLBIGS-PLNGDYQASA-N
SMILES
COC(=O)C(C)=C/C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4350-1.4390 @ 20?C
Form
Liquid

Description

It is employed as a perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a perfuming agent in cosmetics industry.

Solubility
Soluble in alcohol, water 3813 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text