Thermo Scientific Chemicals

1-Fluoro-4-iodobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A12008.09
10 g, Each
Thermo Scientific Chemicals

1-Fluoro-4-iodobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A12008.09
10 g, Each
Quantity
Catalog number: A12008.09
also known as A12008-09
Price (USD)
Price: 34.70
Online price: 29.65
Your price:
Quantity
-

Chemical Identifiers

CAS
352-34-1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5815-1.5845 @ 20?C

Description

4-Fluoroiodobenzene was used in the preparation of fluorotriphenylene derivatives. 4-[F] Fluoroiodobenzene is a versatile building block in 18F radiochemistry and is used in various transition metal-mediated C-C and C-N cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoroiodobenzene was used in the preparation of fluorotriphenylene derivatives. 4-[F] Fluoroiodobenzene is a versatile building block in 18F radiochemistry and is used in various transition metal-mediated C-C and C-N cross-coupling reactions.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool dry place. Keep container tightly sealed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text