Thermo Scientific Chemicals

4-Nitrocatechol, 98+%, Thermo Scientific Chemicals

Catalog number: A12122.22
100 g, Each
Thermo Scientific Chemicals

4-Nitrocatechol, 98+%, Thermo Scientific Chemicals

Catalog number: A12122.22
100 g, Each
Quantity
Catalog number: A12122.22
also known as A12122-22
Price (USD)
Price: 830.00
Online price: 724.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3316-09-4
IUPAC Name
4-nitrobenzene-1,2-diol
Molecular Formula
C6H5NO4
InChI Key
XJNPNXSISMKQEX-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1O)[N+]([O-])=O
Melting Point (clear melt)
172.0-178.0?C
Appearance (Color)
Pale yellow to yellow to brown
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (HPLC)
≥98.0%
Form
Crystals or powder or crystalline powder

Description

4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.

Solubility
Soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text