Thermo Scientific Chemicals

Maleic anhydride, 98+%, Thermo Scientific Chemicals

Catalog number: A12178.0B
1000 g, Each
Thermo Scientific Chemicals

Maleic anhydride, 98+%, Thermo Scientific Chemicals

Catalog number: A12178.0B
1000 g, Each
Quantity
Catalog number: A12178.0B
also known as A12178-0B
Price (USD)
Price: 35.60
Online price: 30.65
Your price:
Quantity
-

Chemical Identifiers

CAS
108-31-6
IUPAC Name
2,5-dihydrofuran-2,5-dione
Molecular Formula
C4H2O3
InChI Key
FPYJFEHAWHCUMM-UHFFFAOYSA-N
SMILES
O=C1OC(=O)C=C1
Appearance (Color)
White
Form
Briquettes or flakes or pellets
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.5%
Free acid (titration)
≤0.5% (as maleic acid)
Identification (FTIR)
Conforms

Description

Maleic anhydride is used in the preparation of unsaturated polyester resins, 4-butanediol, fumairc acid, tetrahydrofuran and gamma-butyrolactone. It is also used as lubricating oil additives, dispersants, artificial sweeteners, flavor enhancements, floor polishes and corrosion inhibitors. It finds application in personal care products like hair sprays. Further, it is used in water treatment chemicals, detergents, pharmaceuticals and copolymers. It is an active substrate for Diels-Alder reactions and dimerizes to form cyclobutane tetracarboxylic dianhydride .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maleic anhydride is used in the preparation of unsaturated polyester resins, 4-butanediol, fumairc acid, tetrahydrofuran and gamma-butyrolactone. It is also used as lubricating oil additives, dispersants, artificial sweeteners, flavor enhancements, floor polishes and corrosion inhibitors. It finds application in personal care products like hair sprays. Further, it is used in water treatment chemicals, detergents, pharmaceuticals and copolymers. It is an active substrate for Diels-Alder reactions and dimerizes to form cyclobutane tetracarboxylic dianhydride .

Solubility
Soluble in water, acetone and dioxane. Slightly soluble in methanol, chloroform and benzene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, strong reducing agents, alkali metals and amines.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text