Thermo Scientific Chemicals

1-Benzyl-4-piperidone, 98+%, Thermo Scientific Chemicals

Catalog number: A12390.14
25 g, Each
Thermo Scientific Chemicals

1-Benzyl-4-piperidone, 98+%, Thermo Scientific Chemicals

Catalog number: A12390.14
25 g, Each
Quantity
Catalog number: A12390.14
also known as A12390-14
Price (USD)
Price: 33.70
Online price: 28.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3612-20-2
IUPAC Name
1-benzylpiperidin-4-one
Molecular Formula
C12H15NO
InChI Key
SJZKULRDWHPHGG-UHFFFAOYSA-N
SMILES
O=C1CCN(CC2=CC=CC=C2)CC1
Identification (FTIR)
Conforms (non-U.S. specification)
Refractive Index
1.5385-1.5425 @ 20°C (non-U.S. specification)
Appearance (Color)
Clear pale yellow to yellow
Form
Liquid
Assay (GC)
≥98.0% (non-U.S. specification)

Description

1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.

Solubility
Soluble in water (12 g/100ml at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text