Thermo Scientific Chemicals

Diethyl diallylmalonate, 97+%, Thermo Scientific Chemicals

Catalog number: A12460.09
10 g, Each
Thermo Scientific Chemicals

Diethyl diallylmalonate, 97+%, Thermo Scientific Chemicals

Catalog number: A12460.09
10 g, Each
Quantity
Catalog number: A12460.09
also known as A12460-09
Price (USD)
Price: 72.50
Online price: 61.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3195-24-2
IUPAC Name
1,3-diethyl 2,2-bis(prop-2-en-1-yl)propanedioate
Molecular Formula
C13H20O4
InChI Key
LYUUVYQGUMRKOV-UHFFFAOYSA-N
SMILES
CCOC(=O)C(CC=C)(CC=C)C(=O)OCC
Appearance (Color)
Clear colorless
Refractive Index
1.4440-1.4490 @ 20?C
Assay (GC)
≥97.0%
Form
Liquid

Description

Diethyl diallylmalonate is the starting material for enantioselective synthesis of carbocyclic nucleoside analogues. It is also used as Low catalyst loading in ring-closing metathesis reaction. Cationic nickel (with monodentate phosphoramidites and Wilke?s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl diallylmalonate is the starting material for enantioselective synthesis of carbocyclic nucleoside analogues. It is also used as Low catalyst loading in ring-closing metathesis reaction. Cationic nickel (with monodentate phosphoramidites and Wilke′s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported.

Solubility
Sparingly Soluble in water. (0.72 g/L) (25°C)

Notes
Store under cool dry place. Ensure good ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text