Bismuth carbonate oxide
Bismuth carbonate oxide
Bismuth carbonate oxide
Thermo Scientific Chemicals

Bismuth carbonate oxide

CAS: 5892-10-4 | CBi2O5 | 509.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A12483.0B
also known as A12483-0B
Price (USD)
282.65
Special offer
Online exclusive
Ends: 30-Jun-2026
332.00
Save 49.35 (15%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
282.65
Special offer
Online exclusive
Ends: 30-Jun-2026
332.00
Save 49.35 (15%)
Each
Chemical Identifiers
CAS5892-10-4
IUPAC Name({[(oxobismuthanyl)oxy]carbonyl}oxy)bismuthanone
Molecular FormulaCBi2O5
InChI KeyMGLUJXPJRXTKJM-UHFFFAOYSA-L
SMILESO=[Bi]OC(=O)O[Bi]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤ 1.0%
Formpowder
Appearance (Color)White to pale cream or pale yellow
Complexometric TitrationBismuth Bi content (dry weight): 80.0 - 82.5% (theory 81.96%)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only