Thermo Scientific Chemicals

trans-4-Methoxy-beta-nitrostyrene, 98%, Thermo Scientific Chemicals

Catalog number: A12745.06
5 g, Each
Thermo Scientific Chemicals

trans-4-Methoxy-beta-nitrostyrene, 98%, Thermo Scientific Chemicals

Catalog number: A12745.06
5 g, Each
Quantity
Catalog number: A12745.06
also known as A12745-06
Price (USD)
Price: 90.80
Online price: 77.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5576-97-6
IUPAC Name
1-methoxy-4-(2-nitroethenyl)benzene
Molecular Formula
C9H9NO3
InChI Key
JKQUXSHVQGBODD-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C[N+]([O-])=O)C=C1
Appearance (Color)
Yellow
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
84.5-90.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text