Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol, 99+%, Thermo Scientific Chemicals

Catalog number: A12747.14
25 g, Each
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol, 99+%, Thermo Scientific Chemicals

Catalog number: A12747.14
25 g, Each
Quantity
Catalog number: A12747.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
920-66-1
IUPAC Name
1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular Formula
C3H2F6O
InChI Key
BYEAHWXPCBROCE-UHFFFAOYSA-N
SMILES
OC(C(F)(F)F)C(F)(F)F
Comment
Material sourced in UK and US
Identification (FTIR)
Conforms (UK-sourced material)
Appearance (Color)
Clear colorless
Assay (GC)
≥99.0% (UK-sourced material)
Refractive Index
<1.3000 @ 20?C (UK-sourced material)

Description

1,1,1,3,3,3-Hexafluoro-2-propanol is used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials. It plays an important role in the Friedel-Crafts reactions due to its polarity and high ionizing power. It finds use in sample preparation for GCMS. It also catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide. It is used as a solvent for many reactions and to dissolve peptides. It is also employed to produce high-end chemicals like fluorinated surfactants, fluorinated emulsifier and fluorinated medicine. It acts as a cleaner in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,3,3,3-Hexafluoro-2-propanol is used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials. It plays an important role in the Friedel-Crafts reactions due to its polarity and high ionizing power. It finds use in sample preparation for GCMS. It also catalyzes the epoxidation of cyclooctene and 1-octene with hydrogen peroxide. It is used as a solvent for many reactions and to dissolve peptides. It is also employed to produce high-end chemicals like fluorinated surfactants, fluorinated emulsifier and fluorinated medicine. It acts as a cleaner in electronic industry.

Solubility
Miscible with water, chloroform, ether and acetone.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases, aluminum, zinc and alkali metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text