Thermo Scientific Chemicals

2-Hydroxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A12971.03
1 g, Each
Thermo Scientific Chemicals

2-Hydroxy-4-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A12971.03
1 g, Each
Quantity
Catalog number: A12971.03
also known as A12971-03
Price (USD)
Price: 23.60
Online price: 19.65
Your price:
Quantity
-

Chemical Identifiers

CAS
673-22-3
IUPAC Name
2-hydroxy-4-methoxybenzaldehyde
Molecular Formula
C8H8O3
InChI Key
WZUODJNEIXSNEU-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=O)C(O)=C1
Appearance (Color)
Pale cream to pale brown or pale pink
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
39.0-46.0?C
Assay (GC)
≥97.5%

Description

It is employed in the synthesis of Schiff base ligand. It is applied as a reactant in the synthesis of LPA1R antagonists used in the inhibition of LPA-induced proliferation and contraction of normal human lung fibroblasts. Also used in the synthesis of tyrosine kinase 6 proteinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of Schiff base ligand. It is applied as a reactant in the synthesis of LPA1R antagonists used in the inhibition of LPA-induced proliferation and contraction of normal human lung fibroblasts. Also used in the synthesis of tyrosine kinase 6 proteinase inhibitors.

Solubility
Solubility in methanol is almost transparent. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents, bases and air/oxygen. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text