Thermo Scientific Chemicals

4-Bromomandelic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12978.22
100 g, Each
Thermo Scientific Chemicals

4-Bromomandelic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12978.22
100 g, Each
Quantity
Catalog number: A12978.22
also known as A12978-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
6940-50-7
IUPAC Name
2-(4-bromophenyl)-2-hydroxyacetic acid
Molecular Formula
C8H7BrO3
InChI Key
BHZBRPQOYFDTAB-UHFFFAOYNA-N
SMILES
OC(C(O)=O)C1=CC=C(Br)C=C1
Melting Point (clear melt)
114.0-120.0?C
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (GC)
≥98.0%

Description

4-Bromomandelic acid is used as a pharmaceutical intermediate. 4-bromomandelic acid is mainly used as an analytical reagent for zirconium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromomandelic acid is used as a pharmaceutical intermediate. 4-bromomandelic acid is mainly used as an analytical reagent for zirconium.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text