Thermo Scientific Chemicals

Methyl 1-methylpyrrole-2-carboxylate, 99%, Thermo Scientific Chemicals

Catalog number: A13095.06
5 g, Each
Thermo Scientific Chemicals

Methyl 1-methylpyrrole-2-carboxylate, 99%, Thermo Scientific Chemicals

Catalog number: A13095.06
5 g, Each
Quantity
Catalog number: A13095.06
also known as A13095-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
37619-24-2
IUPAC Name
methyl 1-methyl-1H-pyrrole-2-carboxylate
Molecular Formula
C7H9NO2
InChI Key
APHVGKYWHWFAQV-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CN1C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5195-1.5225 @ 20?C

Description

Methyl 1-methylpyrrole-2-carboxylate is used as chemical and organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1-methylpyrrole-2-carboxylate is used as chemical and organic Intermediates.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text