Thermo Scientific Chemicals

Bis(2-chloroethyl)amine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13118.30
250 g, Each
Thermo Scientific Chemicals

Bis(2-chloroethyl)amine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A13118.30
250 g, Each
Quantity
Catalog number: A13118.30
also known as A13118-30
Price (USD)
Price: 81.10
Your price:
Quantity
-

Chemical Identifiers

CAS
821-48-7
IUPAC Name
bis(2-chloroethyl)azanium chloride
Molecular Formula
C4H10Cl3N
InChI Key
YMDZDFSUDFLGMX-UHFFFAOYSA-N
SMILES
[Cl-].ClCC[NH2+]CCCl
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder or flakes
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
212.0-220.0?C

Description

Bis(2-chloroethyl)amine hydrochloride is used as an intermediate in chemicals synthesis and building block for piperazine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-chloroethyl)amine hydrochloride is used as an intermediate in chemicals synthesis and building block for piperazine derivatives.

Solubility
Partly miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text