Thermo Scientific Chemicals

2-Hydroxyisobutyric acid, 99% (dry wt.), water <2%, Thermo Scientific Chemicals

Catalog number: A13146.22
100 g, Each
Thermo Scientific Chemicals

2-Hydroxyisobutyric acid, 99% (dry wt.), water <2%, Thermo Scientific Chemicals

Catalog number: A13146.22
100 g, Each
Quantity
Catalog number: A13146.22
also known as A13146-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
594-61-6
IUPAC Name
2-hydroxy-2-methylpropanoic acid
Molecular Formula
C4H8O3
InChI Key
BWLBGMIXKSTLSX-UHFFFAOYSA-N
SMILES
CC(C)(O)C(O)=O
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (dry wt. basis)
Assay (Silylated GC)
≥98.5%
Water Content (Karl Fischer Titration)
≤2.0%
Melting Point (clear melt)
75-81?C

Description

2-Hydroxyisobutyric acid is used as an electrolyte in the separation of the metal ions by on-line cyclic voltammetry. It is also used as a building block for the synthesis of polymers. Its conjugate base, 2-hydroxybutyrate is utilized to catabolize L-threonine as well as synthesize glutathione. Further, it is used as an indicator for the early detection insulin resistance in non-diabetic subjects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxyisobutyric acid is used as an electrolyte in the separation of the metal ions by on-line cyclic voltammetry. It is also used as a building block for the synthesis of polymers. Its conjugate base, 2-hydroxybutyrate is utilized to catabolize L-threonine as well as synthesize glutathione. Further, it is used as an indicator for the early detection insulin resistance in non-diabetic subjects.

Solubility
Soluble in water, ether, alcohol, methanol and hot benzene.

Notes
Incompatible with oxidizing agents, bases and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text