Thermo Scientific Chemicals

6-Methyl-5-hepten-2-one, 98%, Thermo Scientific Chemicals

Catalog number: A13335.22
100 g, Each
Thermo Scientific Chemicals

6-Methyl-5-hepten-2-one, 98%, Thermo Scientific Chemicals

Catalog number: A13335.22
100 g, Each
Quantity
Catalog number: A13335.22
also known as A13335-22
Price (USD)
Price: 35.10
Online price: 29.65
Your price:
Quantity
-

Chemical Identifiers

CAS
110-93-0
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4380-1.4420 @ 20?C

Description

6-Methyl-hepten-2-one is used in the synthesis of thyrsiferyl 23-Acetate which acts as an anti-leukemic inducer of apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-hepten-2-one is used in the synthesis of thyrsiferyl 23-Acetate which acts as an anti-leukemic inducer of apoptosis.

Solubility
Soluble in methanol and chloroform.

Notes
Store in cool, dry place in tightly closed container.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text