6-Methylheptane-2,4-dione, 98+%
6-Methylheptane-2,4-dione, 98+%
6-Methylheptane-2,4-dione, 98+%
Thermo Scientific Chemicals

6-Methylheptane-2,4-dione, 98+%

CAS: 3002-23-1 | C8H14O2 | 142.198 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number A13373.18
also known as A13373-18
Price (USD)
306.65
Special offer
Online exclusive
Ends: 30-Jun-2026
360.00
Save 53.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
306.65
Special offer
Online exclusive
Ends: 30-Jun-2026
360.00
Save 53.35 (15%)
Each
Chemical Identifiers
CAS3002-23-1
IUPAC Name6-methylheptane-2,4-dione
Molecular FormulaC8H14O2
InChI KeyIGMOYJSFRIASIE-UHFFFAOYSA-N
SMILESCC(C)CC(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.0%
Refractive Index1.4550-1.4590 @ 20?C
Identification (FTIR)Conforms
6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylheptane-2,4-dione, is used as a multipurpose intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceutical industry

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from Strong oxidizing agents.
RUO – Research Use Only

General References:

  1. BK Kaymakçıoğlu.; S Rollas. Synthesis, characterization and evaluation of antituberculosis activity of some hydrazones. Il Farmaco. 2002, 57,(7), 595-599.
  2. P Weyerstahl.; U Splittgerber. Constituents of the leaf essential oil of Hypericum perforatum L. from India. Flavour and fragrance. 1995, 10,(6), 365-370.
  3. Chelating ligand.