1,3-Diphenyl-1,3-propanedione, 98+%
1,3-Diphenyl-1,3-propanedione, 98+%
1,3-Diphenyl-1,3-propanedione, 98+%
Thermo Scientific Chemicals

1,3-Diphenyl-1,3-propanedione, 98+%

CAS: 120-46-7 | C15H12O2 | 224.259 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13450.36
also known as A13450-36
Price (USD)
540.65
Online Exclusive
601.00
Save 60.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
540.65
Online Exclusive
601.00
Save 60.35 (10%)
Each
Chemical Identifiers
CAS120-46-7
IUPAC Name1,3-diphenylpropane-1,3-dione
Molecular FormulaC15H12O2
InChI KeyNZZIMKJIVMHWJC-UHFFFAOYSA-N
SMILESO=C(CC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting point75-80°C

1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer & antitumour agent. It is used in perfumery and as well as in organic synthesis.

Solubility
It is Soluble in ether, chloroform, and aqueous sodium hydroxide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Acid anhydrides, Halogens, halogen acids. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. B Kaitner.; E Mestrovic. Structure of a new crystal modification of 1, 3-diphenyl-1, 3-propanedione. Acta Crystallographica Section C: Crystal.1993, C49,1523-1525.
  2. Chelating ligand, e.g. for metal extraction. Reagent for uranium: Anal. Chim. Acta, 108, 437 (1979).