Thermo Scientific Chemicals

p-Chloranil, 97%, Thermo Scientific Chemicals

Catalog number: A13495.22
100 g, Each
Thermo Scientific Chemicals

p-Chloranil, 97%, Thermo Scientific Chemicals

Catalog number: A13495.22
100 g, Each
Quantity
Catalog number: A13495.22
also known as A13495-22
Price (USD)
Price: 119.00
Online price: 101.65
Your price:
Quantity
-

Chemical Identifiers

CAS
118-75-2
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
Yellow to yellow-green
Assay (HPLC)
≥96.0%

Description

p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.

Solubility
Soluble in ether and acetone. Slightly soluble in chloroform, dimethylformamide, solvent naphtha, benzene, carbon disulfide and carbon tetrachloride. Insoluble in water, cold petroleum ether, methanol, dibutyl phthalate and cold alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text