Thermo Scientific Chemicals

trans-Cinnamic acid, 99+%, Thermo Scientific Chemicals

Catalog number: A13538.36
500 g, Each
Thermo Scientific Chemicals

trans-Cinnamic acid, 99+%, Thermo Scientific Chemicals

Catalog number: A13538.36
500 g, Each
Quantity
Catalog number: A13538.36
also known as A13538-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
140-10-3
IUPAC Name
(2E)-3-phenylprop-2-enoic acid
Molecular Formula
C9H8O2
InChI Key
WBYWAXJHAXSJNI-VOTSOKGWSA-N
SMILES
OC(=O)\C=C\C1=CC=CC=C1
Assay from Supplier's CofA
≥99.0%
Residue on ignition
≤0.5%
Melting Point
132-136°C
Appearance (Color)
White to pale cream
Form
Powder or crystals or crystalline powder

Description

trans-Cinnamic acid is used in flavors, synthetic indigo and pharmaceuticals. It is involved in the production of methyl, ethyl and benzyl esters, which is used in the perfume industry. It serves as a precursor to the sweetener aspartame through enzyme-catalyzed amination to phenylalanine. It is a self-inhibitor produced by fungal spores to prevent germination. In addition, it is used to establish phenolic compounds by liquid chromatography, ultraviolet and mass spectrometry. It is utilized as a potential agent, thereby preventing lung tumor cells from metastasizing. Further, it induces intracellular release of calcium ions from the vacuole to the cytoplasm in order to trigger phytotoxicity in cucumber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Cinnamic acid is used in flavors, synthetic indigo and pharmaceuticals. It is involved in the production of methyl, ethyl and benzyl esters, which is used in the perfume industry. It serves as a precursor to the sweetener aspartame through enzyme-catalyzed amination to phenylalanine. It is a self-inhibitor produced by fungal spores to prevent germination. In addition, it is used to establish phenolic compounds by liquid chromatography, ultraviolet and mass spectrometry. It is utilized as a potential agent, thereby preventing lung tumor cells from metastasizing. Further, it induces intracellular release of calcium ions from the vacuole to the cytoplasm in order to trigger phytotoxicity in cucumber.

Solubility
Soluble in many organic solvents. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text