Thermo Scientific Chemicals

1,1,1-Trifluoroacetone, 95%, Thermo Scientific Chemicals

Catalog number: A13556.22
100 g, Each
Thermo Scientific Chemicals

1,1,1-Trifluoroacetone, 95%, Thermo Scientific Chemicals

Catalog number: A13556.22
100 g, Each
Quantity
Catalog number: A13556.22
also known as A13556-22
Price (USD)
Price: 351.00
Online price: 298.65
Your price:
Quantity
-

Chemical Identifiers

CAS
421-50-1
IUPAC Name
1,1,1-trifluoropropan-2-one
Molecular Formula
C3H3F3O
InChI Key
FHUDAMLDXFJHJE-UHFFFAOYSA-N
SMILES
CC(=O)C(F)(F)F
Appearance (Color)
Clear colorless to pale yellow to yellow to orange
Assay (GC)
≥94.0%
Refractive Index
<1.3000 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

1,1,1-Trifluoroacetone is used in a synthesis of 2-trifluoromethyl-7-azaindoles starting with 2,6-dihalopyridines. The derived chiral imine was used to prepare enantiopure α-trifluoromethyl alanines and diamines via a Strecker reaction followed by either nitrile hydrolysis or reduction. And also used as a useful molecule in synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trifluoroacetone is used in a synthesis of 2-trifluoromethyl-7-azaindoles starting with 2,6-dihalopyridines. The derived chiral imine was used to prepare enantiopure α-trifluoromethyl alanines and diamines via a Strecker reaction followed by either nitrile hydrolysis or reduction. And also used as a useful molecule in synthesis reactions.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text