Thermo Scientific Chemicals

1-Chloro-4-fluorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A13690.14
25 g, Each
Thermo Scientific Chemicals

1-Chloro-4-fluorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A13690.14
25 g, Each
Quantity
Catalog number: A13690.14
also known as A13690-14
Price (USD)
Price: 53.40
Online price: 45.65
Your price:
Quantity
-

Chemical Identifiers

CAS
352-33-0
IUPAC Name
1-chloro-4-fluorobenzene
Molecular Formula
C6H4ClF
InChI Key
RJCGZNCCVKIBHO-UHFFFAOYSA-N
SMILES
FC1=CC=C(Cl)C=C1
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Refractive Index
1.4930-1.4970 @ 20?C

Description

1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4'-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 '-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4′-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 ′-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text