Thermo Scientific Chemicals

Diethyl iodomethylphosphonate, 98+%, Thermo Scientific Chemicals

Catalog number: A13697.22
100 g, Each
Thermo Scientific Chemicals

Diethyl iodomethylphosphonate, 98+%, Thermo Scientific Chemicals

Catalog number: A13697.22
100 g, Each
Quantity
Catalog number: A13697.22
also known as A13697-22
Price (USD)
Price: 1,202.00
Online price: 1,050.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10419-77-9
Assay (GC)
≥98.0%
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4970-1.5010 @ 20?C

Description

Diethyl iodomethylphosphonate is used in the synthesis of H2N-Tyr(tBu)-vinyl sulfone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl iodomethylphosphonate is used in the synthesis of H2N-Tyr(tBu)-vinyl sulfone.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text