Thermo Scientific Chemicals

N-Benzylmethylamine, 97%, Thermo Scientific Chemicals

Catalog number: A13746.AK
250 mL, Each
Thermo Scientific Chemicals

N-Benzylmethylamine, 97%, Thermo Scientific Chemicals

Catalog number: A13746.AK
250 mL, Each
Quantity
Catalog number: A13746.AK
also known as A13746-AK
Price (USD)
Price: 37.00
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
103-67-3
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.5195-1.5245 @ 20°C
Form
Liquid

Description

N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.

Solubility
Soluble in water (65 g/L at 20°C).

Notes
Air Sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text