Thermo Scientific Chemicals

4-Aminobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A13794.09
10 g, Each
Thermo Scientific Chemicals

4-Aminobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A13794.09
10 g, Each
Quantity
Catalog number: A13794.09
also known as A13794-09
Price (USD)
Price: 37.50
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
873-74-5
IUPAC Name
4-aminobenzonitrile
Molecular Formula
C7H6N2
InChI Key
YBAZINRZQSAIAY-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1)C#N
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale cream to cream to yellow to orange to brown to grey
Assay (GC)
≥97.5%
Melting Point (clear melt)
82.0-88.0?C

Description

4-Aminobenzonitrile is used as an amino substituted benzonitrile with hypotensive activity and as a radioprotective agent. It is also used as a derivatization reagent in the capillary zone electrophoretic analysis of aldoses, ketoses and uronic acid. Further, it is used in the preparation of methacrylic monomers containing pendant azobenzene structures and polythiophenes containing an azobenzene moiety in the side-chain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzonitrile is used as an amino substituted benzonitrile with hypotensive activity and as a radioprotective agent. It is also used as a derivatization reagent in the capillary zone electrophoretic analysis of aldoses, ketoses and uronic acid. Further, it is used in the preparation of methacrylic monomers containing pendant azobenzene structures and polythiophenes containing an azobenzene moiety in the side-chain.

Solubility
Soluble in ethyl acetate, dichloromethane and chloroform. Insoluble in water.

Notes
Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text