Thermo Scientific Chemicals

Benzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14062.0B
1000 g, Each
Thermo Scientific Chemicals

Benzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: A14062.0B
1000 g, Each
Quantity
Catalog number: A14062.0B
also known as A14062-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65-85-0
IUPAC Name
benzoic acid
Molecular Formula
C7H6O2
InChI Key
WPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or flakes
Identification (FTIR)
Conforms
Melting Point (clear melt)
120.0-126.0?C
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

Benzoic acid is an important precursor for the preparation of many other organic substances viz. benzoyl chloride, benylbenzoate, phenol, sodium benzoate, benzyl alcohol as well as benzoate plasticizers such as glycol-, diethhyleneglycol- and triethyleneglycol esters. Benzoic acid and its salts are used as food preservatives. It is involved in rubber polymerization as an activator and a retardant. It is the main component of benzoin resin and is a constituent of Whitfield’s ointment, which is used for the treatment of fungal skin diseases such as tinea, ringworm and athlete’s foot. It is widely used in cosmetics, dyes, plastics and insect repellents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in acetone, benzene, chloroform, alcohol, diethyl ether, hexane and liquid ammonia. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, reducing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text