Thermo Scientific Chemicals

4-Fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14129.22
100 g, Each
Thermo Scientific Chemicals

4-Fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A14129.22
100 g, Each
Quantity
Catalog number: A14129.22
also known as A14129-22
Price (USD)
Price: 90.80
Online price: 77.65
Your price:
Quantity
-

Chemical Identifiers

CAS
456-22-4
IUPAC Name
4-fluorobenzoic acid
Molecular Formula
C7H5FO2
InChI Key
BBYDXOIZLAWGSL-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(F)C=C1
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Form
Powder
Appearance (Color)
White to cream
Identification (FTIR)
Conforms

Description

4-Fluorobenzoic acid is used as an intermediate for agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzoic acid is used as an intermediate for agrochemicals and pharmaceuticals.

Solubility
Soluble in alcohol, hot water, methanol, and ether. Very slightly soluble in cold water.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text