Thermo Scientific Chemicals

1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A14140.09
10 g, Each
Thermo Scientific Chemicals

1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A14140.09
10 g, Each
Quantity
Catalog number: A14140.09
also known as A14140-09
Price (USD)
Price: 79.50
Online price: 67.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5144-89-8
IUPAC Name
1,10-phenanthroline hydrate
Molecular Formula
C12H10N2O
InChI Key
PPQJCISYYXZCAE-UHFFFAOYSA-N
SMILES
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Assay from Suppliers CofA
≥99.0%
Appearance (Color)
White to cream
Melting Point (clear melt)
95-103°C
Form
Crystalline powder or needles

Description

1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.

Solubility
Soluble in ethanol, acetone and methanol. Slightly soluble in water and benzene.

Notes
Working solutions of 1,10-Phenanthroline monohydrate may stable for a few days at refrigerated temperatures. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text