Thermo Scientific Chemicals

Isododecane, mixture of isomers, tech. 80%, Thermo Scientific Chemicals

Catalog number: A14235.AP
500 mL, Each
Thermo Scientific Chemicals

Isododecane, mixture of isomers, tech. 80%, Thermo Scientific Chemicals

Catalog number: A14235.AP
500 mL, Each
Quantity
Catalog number: A14235.AP
Price (USD)
Price: 82.40
Online price: 70.65
Your price:
Quantity
-

Chemical Identifiers

CAS
31807-55-3
IUPAC Name
2-methylundecane
Molecular Formula
C12H26
InChI Key
GTJOHISYCKPIMT-UHFFFAOYSA-N
SMILES
CCCCCCCCCC(C)C
Appearance (Color)
Clear colorless
Assay (GC)
≥78.0% (major isomer)
Assay (GC)
≥92.0% (mixture of isomers)
Refractive Index
1.4180-1.4240 @ 20?C
Form
Liquid

Description

Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isododecane, mixture of isomer acts as a reaction medium used for polymerization reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text