Thermo Scientific Chemicals

Folic acid dihydrate, 97%, Thermo Scientific Chemicals

Catalog number: A14300.14
25 g, Each
Thermo Scientific Chemicals

Folic acid dihydrate, 97%, Thermo Scientific Chemicals

Catalog number: A14300.14
25 g, Each
Quantity
Catalog number: A14300.14
also known as A14300-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
75708-92-8
IUPAC Name
2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid dihydrate
Molecular Formula
C19H23N7O8
InChI Key
ODYNNYOEHBJUQP-UHFFFAOYNA-N
SMILES
O.O.NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)NC(CCC(O)=O)C(O)=O)=CN=C2N1
Appearance (Color)
Yellow to orange
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0% (UK sourced material)
Assay from Supplier's CofA
≥96.0 to ≤104.0% (US sourced material)
Comment
Material sourced in UK and US
Water Content (Karl Fischer Titration)
≤8.5% (UK and US sourced material)

Description

Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid dehydrate is used to form non-covalent complexes with polyethylenimine. It is used for the delivery of genes, which finds application in gene therapy. It is a B-complex vitamin, which is necessary for the body to produce red blood cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid dehydrate is used to form non-covalent complexes with polyethylenimine. It is used for the delivery of genes, which finds application in gene therapy. It is a B-complex vitamin, which is necessary for the body to produce red blood cells.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text